Order Entry
Denmark
ContactUsLinkComponent
TCLP Acid mix (6 components)
TCLP Acid mix (6 components)
Catalog # REST31027
Supplier:  Restek
CAS Number:  
undefined
TCLP Acid mix (6 components)
Catalog # REST31027
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml in P and T methanol
  • Pk:
    1 mL
  • Description:
    TCLP Acid mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

TCLP acid mix contains 2-methylphenol (o-cresol), 3-methylphenol (m-cresol), 4-methylphenol (p-cresol), pentachlorophenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.