Order Entry
Denmark
ContactUsLinkComponent
Environmental standard, minnesota Ag list 1 pesticides mix A (16 components)
Environmental standard, minnesota Ag list 1 pesticides mix A (16 components)
  REST32406
 :  Restek
 :  
undefined
Environmental standard, minnesota Ag list 1 pesticides mix A (16 components)
  REST32406
 :  Restek
 :  

 

  • Concentration:
    200 ppm each in acetone
  • Pk:
    1 mL
  • Description:
    Minnesota Ag list 1 pesticides mix A
  • Pk Info:
    1 ml/ampoule

 

 

Contains: Acetochlor, alachlor, atrazine, cyanazine (bladex), desethylatrazine, desisopropylatrazine, dimethenamid, ethalfluralin, metolachlor, metribuzin, pendimethalin, prometon, propachlor, propazine, simazine, trifluralin.

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.