Order Entry
Denmark
ContactUsLinkComponent
DHA isoparaffins standard
DHA isoparaffins standard
Catalog # REST30726
Supplier:  Restek
CAS Number:  
undefined
DHA isoparaffins standard
Catalog # REST30726
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    0,15 mL
  • Description:
    DHA isoparaffins standard, 0.15 ml in autosampler vial

Specifications

About this item

This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.

It contain 33 components: 3,3-Diethylpentane (1,914%), 2,3-Dimethylbutane (2,352%), 2,3-Dimethylheptane (1,16%), 2,5-Dimethylheptane (3,86%), 3,3-Dimethylheptane (1,765%), 3,4-Dimethylheptane (0,839%), 3,5-Dimethylheptane (2,344%), 2,2-Dimethylhexane (0,786%), 2,4-Dimethylhexane (1,454%), 2,5-Dimethylhexane (5,854%), 2,2-Dimethyloctane (2,095%), 3,3-Dimethyloctane (0,804%), 2,2-Dimethylpentane (2,438%), 2,3-Dimethylpentane (4,668%), 2,4-Dimethylpentane (5,931%), 3,3-Dimethylpentane (2,305%), 3-Ethylhexane (0,307%).

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.

Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)