Order Entry
Denmark
ContactUsLinkComponent
DHA paraffins standard
DHA paraffins standard
Catalog # REST30725
Supplier:  Restek
CAS Number:  
undefined
DHA paraffins standard
Catalog # REST30725
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    0,15 mL
  • Description:
    DHA paraffins standard
  • Pk Info:
    0.15 ml in autosampler vial

Specifications

About this item

This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration to help our customers optimize production and maximize profitability—while fulfilling requirements such as ASTM Methods D6729, D6730, and D6733.

It contain 11 components: N-Decane, 9,134%, N-Dodecane, 9,089%, N-Heptane, 9,066%, N-Hexane, 9,097%, N-Nonane, 9,086%, N-Octane, 9,054%, N-Pentadecane, 9,123%, N-Pentane, 9,094%, N-Tetradecane, 9,128%, N-Tridecane, 9,064%, N-Undecane, 9,064%.