SOM01.1 VOA ketone deuterated monitoring compound
REST30625
: Restek
:
- Concentration:500 µg/ml each in deuterium oxide (D₂O)
- Pk:1 Ampoul
- Description:SOM01.1 VOA Ketone Deuterated Monitoring Compounds
- Pk Info:1 ml/ampoule
This ketone deuterated monitoring compound contains two components: 2-butanone-d5 and 2-hexanone-d5.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.