Pesticide reference standard, Methoxychlor
Catalog # REST32233
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in methanol
- Pk:1
- CAS No.:72-43-5
- Description:Methoxychlor
- Formula:C₁₆H₁₅Cl₃O₂
- Description:Methoxychlor
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.