Order Entry
Denmark
ContactUsLinkComponent
LC OQ wavelength accuracy standard
LC OQ wavelength accuracy standard
Catalog # REST31053
Supplier:  Restek
CAS Number:  
LC OQ wavelength accuracy standard
Catalog # REST31053
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 Ampoul
  • Description:
    Erbium perchlorate (14017-55-1), 10 mg/ml in water
  • Pk Info:
    5 ml/ampoule
Specifications
About this item
Erbium perchlorate, 14017-55-1