Alachor, reference standard
Alachlor, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32204
Supplier: Restek
Jump to:
Product Details & Documents
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pakn:1
- Beskrivelse:Alachlor, 1000 µg/ml in methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST32204
- Beskrivelse:Alachlor
- CAS-nr:15972-60-8
- Formel:C₁₄H₂₀ClNO₂



