Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Hydrochloric acid fuming 36.5 - 38%, meets analytical specification of BP, Ph. Eur., NF, Fluka™
 
 
 
Hydrochloric acid fuming 36.5 - 38%, meets analytical specification of BP, Ph. Eur., NF, Fluka™
 

 

  • Assay:
    36.5 - 38.0 %
  • Residue on ignition (SO₄):
    Max. 0.002 %
  • Non-volatile matter:
    Max. 0.005 %
  • Arsenic (As):
    Max. 0.00005 %
  • Iron (Fe):
    Max. 0.0001 %
  • Free chlorine (Cl):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 2 ppm
  • Sulfite (SO₃):
    Max. 0.0005 %
  • Free Bromine or Chlorine:
    complying
  • Bromide or Iodide:
    complying
  • Appearance of the substance:
    complying
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying
  • Vapor Density:
    1.3 (vs air)

 

 

Frequently Bought Together