Order Entry
Denmark
ContactUsLinkComponent
Magnesium oxide light, dried 98.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E530, Fluka™
Magnesium oxide light, dried 98.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E530, Fluka™
Certificates
Magnesium oxide light, dried 98.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E530, Fluka™
Certificates

Specification Test Results

  • Assay (calculated to ignited substance):
    98.0 - 100.5 %
  • Loss on ignition:
    Max. 5.0 %
  • In water soluble matter:
    Max. 1.0 %
  • Insoluble in acetic acid:
    Max. 0.1 %
  • Insoluble in HCl:
    Max. 0.1 %
  • Arsenic (As):
    Max. 2 ppm
  • Calcium (Ca):
    Max. 1 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.02 %
  • Lead (Pb):
    Max. 2 ppm
  • Zinc (Zn):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Chloride (Cl):
    Max. 0.05 %
  • Sulfate (SO₄):
    Max. 0.5 %
  • Free alkali + Soluble salts:
    complying
  • Appearance of the solution:
    complying
  • Bulk Density:
    Max. 0.15 g/ml
  • Residual solvents:
    complying
  • Filling Volume:
    ≤0.15 g/mL

Specification Test Results

Related Information

Frequently Bought Together