Order Entry
Denmark
ContactUsLinkComponent
Diammonium hydrogen citrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Irritant
Diammonium hydrogen citrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # 09831-500G
CAS Number:  
Irritant
Diammonium hydrogen citrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Catalog # 09831-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Residue on ignition:
    Max. 0.01 %
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 500 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Oxalate (C₂O₄):
    Max. 500 ppm
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :226.19 g/mol
Merck Index:13,00513
Density:1.480 g/cm³ (20 °C)
EINECS:221-146-3
 :GHS07
Storage Temperature:Ambient
CAS Number:3012-65-5
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068