Order Entry
Denmark
ContactUsLinkComponent
 
 
Ammonium formate ≥97.0% (by titrimetric analysis), Purum p.a.
  09739-100G
 :  
 
Ammonium formate ≥97.0% (by titrimetric analysis), Purum p.a.
  09739-100G
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Identity (IR):
    complying
  • Assay:
    97 - 103 %
  • pH (5 %, 25 °C):
    6.0 - 7.0
  • Colour of the solution:
    complying
  • Turbidity of solution:
    complying
  • Water (Karl Fischer):
    Max. 3 %
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 50 ppm
  • Cobalt (Co):
    Max. 50 ppm
  • Copper (Cu):
    Max. 50 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Potassium (K):
    Max. 200 ppm
  • Sodium (Na):
    Max. 100 ppm
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 50 ppm
  • Zinc (Zn):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 100 ppm

 

 

 :
 :63.06 g/mol
 :13,00525
 :116 °C
 :1.266 g/cm3
 :208-753-9
 :GHS07
 :Ambient
 :540-69-2
 :HCO₂NH₄
 :MFCD00013103