Sodium acetate trihydrate ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Merck Index:13,08642
Melting Pt:58 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
Specification Test Results
- Assay:99.0 - 101.0 %
- Assay (calculated to the dried substance):99.0 - 101.0 %
- Water insoluble matter:Max. 0.005 %
- Loss on drying (130°C):39.0 - 40.5 %
- pH (5 %, 20°C):7.5 - 9.0
- pH (5 %, 25 °C):7.5 - 9.2
- Aluminium (Al):Max. 0.2 ppm
- Arsenic (As):Max. 2 ppm
- Calcium (Ca):Max. 20 ppm
- Cadmium (Cd):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 20 ppm
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 5 ppm
- Phosphate (PO₄):Max. 0.0005 %
- Sulfate (SO₄):Max. 20 ppm
- KMnO₄ reducing matter (as HCOOH):Max. 0.005 %
- Reducing impurities:complying
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together