Order Entry
Denmark
ContactUsLinkComponent
Lead(II) nitrate ≥99.0% ACS, Fluka™
Irritant
Corrosive
Health
Oxidizing
Lead(II) nitrate ≥99.0% ACS, Fluka™
Certificates
Irritant
Corrosive
Health
Oxidizing
Lead(II) nitrate ≥99.0% ACS, Fluka™

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • In diluted HNO₃ insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 50 ppm
  • Copper (Cu):
    Max. 20 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 50 ppm
  • Sodium (Na):
    Max. 200 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Form:
    Solid

Specification Test Results

Related Information

Frequently Bought Together