Order Entry
Denmark
ContactUsLinkComponent
 
Nitric acid ≥70% ACS, Fluka™
 
Nitric acid ≥70% ACS, Fluka™
 

 

  • Environmentally Preferable:

 

 

  • Appearance (color):
    complying
  • Appearance (form):
    complying
  • Assay:
    68.0 - 70.0 %
  • Residue on ignition (SO₄):
    Max. 5 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Heavy metals (as Pb):
    Max. 0.2 ppm
  • Chloride (Cl):
    Max. 0.5 ppm
  • Sulfate (SO₄):
    Max. 1 ppm
  • APHA:
    Max. 10
  • Ignition Residue:
    ≤5 ppm

 

 

Frequently Bought Together