Order Entry
Denmark
ContactUsLinkComponent
Potassium chromate ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Potassium chromate ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Certificates
Potassium chromate ≥99.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    8.6 - 9.8
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.002 %
  • Sodium (Na):
    Max. 0.02 %
  • Lead (Pb):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together