Order Entry
Denmark
ContactUsLinkComponent
 
 
Lithium carbonate ≥99%, Purum, Fluka™
 
 
Lithium carbonate ≥99%, Purum, Fluka™

 

  • Assay:
    Min. 99 %
  • Calcium (Ca):
    Max. 0.03 %
  • Iron (Fe):
    Max. 0.003 %
  • Potassium (K):
    Max. 0.03 %
  • Magnesium (Mg):
    Max. 0.01 %
  • Sodium (Na):
    Max. 0.1 %
  • Heavy metals (as Pb):
    Max. 0.003 %
  • Chloride (Cl):
    Max. 0.02 %
  • Sulfate (SO₄):
    Max. 0.3 %

 

Frequently Bought Together