Order Entry
Denmark
ContactUsLinkComponent
2-Methylbutane ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Flammable
Health
Irritant
2-Methylbutane ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
Flammable
Health
Irritant
2-Methylbutane ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.50 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 240 nm:
    Max. 0.01
  • Absorbance at 220 nm:
    Max. 0.07
  • Absorbance at 210 nm:
    Max. 0.30
  • Absorbance at 192 nm:
    Max. 1.0
  • Vapor Density:
    2.6 (vs air)
  • Autoignition Temperature:
    788 °F

Specification Test Results

Related Information

Frequently Bought Together