Order Entry
Denmark
ContactUsLinkComponent
2-Methylbutane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Irritant
Health
Flammable
2-Methylbutane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # 59070-1L
CAS Number:  
undefined
Irritant
Health
Flammable
2-Methylbutane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # 59070-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.353 - 1.355
  • Water (Karl Fischer):
    Max. 0.05 %
  • Non-volatile matter:
    Max. 0.001 %
  • Identity (NMR):
    complying
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Explosion Limit:
    8.3 %

Specification Test Results

About this item

 :
 :72.15 g/mol
Boiling Pt:28 °C (1.013 hPa)
Melting Pt:-160 °C
Melting Pt:–160 °C
Density:0.62 g/cm³ (20 °C)
Flash Pt:7.6 %(V)
Storage Temperature:Ambient
EINECS:201-142-8
UN:1265
ADR:3,I
 :GHS07
 :GHS08
 :GHS02
CAS Number:78-78-4
Formula:CH₃CH₂CH(CH₃)₂
MDL Number:MFCD00009338