Ammonium acetate ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:13,00495
Melting Pt:113 °C
Density:1.170 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-162-9
Specification Test Results
- Assay:Min. 98 %
- Water insoluble matter:Max. 0.005 %
- Water (Karl Fischer):Max. 2 %
- Sulfated ash:Max. 0.01 %
- pH (5 %, 25 °C):6.7 - 7.3
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0002 %
- Cobalt (Co):Max. 0.0005 %
- Copper (Cu):Max. 0.0002 %
- Chromium (Cr):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.0002 %
- Manganese (Mn):Max. 0.0005 %
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0002 %
- Zinc (Zn):Max. 0.0002 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.0005 %
- Nitrate (NO₃):Max. 0.001 %
- Sulfate (SO₄):Max. 0.001 %
- KMnO₄ reducing matter (as HCOOH):Max. 0.005 %
Specification Test Results
Frequently Bought Together