Jumbo syringes
: Restek
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
- Central port with a Luer lock fitting
- Secondary port with septum nut
- Detachable plunger stem
- Easy sample storage
An adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time.
Product Family Options
- VolumeType