Order Entry
Denmark
ContactUsLinkComponent
Appendix IX mix #2
Appendix IX mix #2
Catalog # REST31806
Supplier:  Restek
CAS Number:  
Appendix IX mix #2
Catalog # REST31806
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 Ampoul
  • Description:
    Appendix IX Mix #2 (32 components),
    1000 µg/ml each in methylene chloride
  • Pk Info:
    1 ml/ampoule
  • Type:
    EPA Appendix IX
Specifications
About this item
Appendix IX mix, contains 32 components.