Order Entry
Denmark
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution, high sensitivity ready-to-use, precipitating
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution, high sensitivity ready-to-use, precipitating
Catalog # J62143.AP
CAS Number:  
3,3',5,5'-Tetramethylbenzidine (TMB) in aqueous solution, high sensitivity ready-to-use, precipitating
Catalog # J62143.AP
CAS Number:  

Specifications

  • Pk:
    500 mL

Specifications

About this item

 :
 :240.35 g/mol
UN:2810
 :GHS08
 :GHS07
Storage Temperature:Refrigerator
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748