PCB congeners
Supplier: Restek
About this item
Certified reference materials.
Manufactured and QC-tested in ISO-accredited labs.
7 Options Available Below
Product Details & Documents
Certified reference materials.
Manufactured and QC-tested in ISO-accredited labs.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- REST32288
- REST32286
- REST32287
- REST32285
- REST32293
- REST32283
- REST32284
- Description:
- PCB congener standard includes 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes 2,2',4,5,5'-pentachlorobiphenyl (BZ #101) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampoule
- PCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoule
- CAS No.:
- 35065-29-3
- 35065-28-2
- 35065-27-1
- 37680-73-2
- 31508-00-6
- 7012-37-5
- 35693-99-3
- Formula:
- C12H3Cl7
- C12H4Cl6
- C12H4Cl6
- C12H5Cl5
- C12H5Cl5
- C12H7Cl3
- C12H6Cl4
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- REST32288PCB congener standard includes 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180) with 10 µg/ml in isooctane, 1 ml/ampoule1 mLLog in for availability1 * 1 mL265,00 krSpecifications:
More
Specifications
Cat. No.REST32288DescriptionPCB congener standard includes 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.35065-29-3FormulaC12H3Cl7 - REST32286PCB congener standard includes 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138) with 10 µg/ml in isooctane, 1 ml/ampoule1Log in for availability1 * 1 ItemsSpecifications:
More
Specifications
Cat. No.REST32286DescriptionPCB congener standard includes 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.35065-28-2FormulaC12H4Cl6 - REST32287PCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampoule1Log in for availability1 * 1 ItemsSpecifications:
More
Specifications
Cat. No.REST32287DescriptionPCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.35065-27-1FormulaC12H4Cl6 - REST32285PCB congener standard includes 2,2',4,5,5'-pentachlorobiphenyl (BZ #101) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,2',4,5,5'-pentachlorobiphenyl (BZ #101) with 10 µg/ml in isooctane, 1 ml/ampoule1Log in for availability1 * 1 ItemsSpecifications:
More
Specifications
Cat. No.REST32285DescriptionPCB congener standard includes 2,2',4,5,5'-pentachlorobiphenyl (BZ #101) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.37680-73-2FormulaC12H5Cl5 - REST32293PCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampoule1 mLLog in for availability1 * 1 mL418,00 krSpecifications:
More
Specifications
Cat. No.REST32293DescriptionPCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.31508-00-6FormulaC12H5Cl5 - REST32283PCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampoule1 mLLog in for availability1 * 1 mLSpecifications:
More
Specifications
Cat. No.REST32283DescriptionPCB congener standard includes 2,4,4'-trichlorobiphenyl (BZ #28) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.7012-37-5FormulaC12H7Cl3 - REST32284PCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoulePCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoule1Log in for availability1 * 1 ItemsSpecifications:
More
Specifications
Cat. No.REST32284DescriptionPCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampouleCAS No.35693-99-3FormulaC12H6Cl4



