Order Entry
Denmark
ContactUsLinkComponent
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (G.C.):
    Min. 99,5 %
  • Identity (IR-spectrum):
    Passes test
  • Density (20º/4º):
    0,964-0,968 g/ml
  • Density (20º/20º):
    Approx. 0,97 g/ml
  • Refractive index (n20/D):
    Approx. 1,403
  • Boiling point:
    Approx. 125ºC
  • Appearance:
    Clear
  • Colour (Hazen):
    Max. 10
  • Acidity:
    Max. 0,001 meq/g
  • Alkalinity:
    Max. 0,0005 meq/g
  • Aluminium (Al):
    Max. 0,00005 %
  • Barium (Ba):
    Max. 0,00001 %
  • Boron (B):
    Max. 0,000002 %
  • Cadmium (Cd):
    Max. 0,000005 %
  • Calcium (Ca):
    Max. 0,00005 %
  • Chromium (Cr):
    Max. 0,000002 %
  • Cobalt (Co):
    Max. 0,000002 %
  • Copper (Cu):
    Max. 0,000002 %
  • Iron (Fe):
    Max. 0,00005 %
  • Lead (Pb):
    Max. 0,00001 %
  • Magnesium (Mg):
    Max. 0,00001 %
  • Manganese (Mn):
    Max. 0,000002 %
  • Nickel (Ni):
    Max. 0,000002 %
  • Tin (Sn):
    Max. 0,00001 %
  • Zinc (Zn):
    Max. 0,00001 %
  • Aldehydes (as CH3CHO):
    Max. 0,003 %
  • Formaldehyde:
    Max. 0,001 %
  • Peroxides (as H2O2):
    Max. 0,002 %
  • Substances darkened by H2SO4:
    Passes test
  • Residue on evaporation:
    Max. 0,002 %
  • Water (K.F.):
    Max. 0,1 %

Specification Test Results

Related Information

Frequently Bought Together