Specifications
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:121776-33-8
- Description:Furilazol
- Environmentally Preferable:
- Melting point:95…100 °C
- Formula:C₁₁H₁₃Cl₂NO₃
- Storage temperature:Ambient
- Description:Furilazol
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results