Order Entry
Denmark
ContactUsLinkComponent
 
Analytical standard, Moxifloxacin hydrochloride, reference material, Honeywell Fluka™
  HONCR1181-50MG
 :  
Analytical standard, Moxifloxacin hydrochloride, reference material, Honeywell Fluka™
  HONCR1181-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    186826-86-8
  • Description:
    Moxifloxacin hydrochloride
  • Melting point:
    233 °C
  • Formula:
    C₂₁H₂₄FN₃O₄· HCl
  • Storage temperature:
    2…8 °C
  • Description:
    Moxifloxacin hydrochloride

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results