Analytical standard, Cyantraniliprole, reference material, Honeywell Fluka™
Catalog # HONCR1180-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:736994-63-1
- Description:Cyantraniliprole
- Environmentally Preferable:
- Melting point:150 °C
- Formula:C₁₉H₁₄BrClN₆O₂
- Storage temperature:Ambient
- Description:Cyantraniliprole
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results