Order Entry
Denmark
ContactUsLinkComponent
 
Analytical standard, (±)-Metoprolol-(isopropyl-d₇) (+)-tartrate salt, reference material, Honeywell Fluka™
Catalog # HONCR1162-10MG
CAS Number:  
Analytical standard, (±)-Metoprolol-(isopropyl-d₇) (+)-tartrate salt, reference material, Honeywell Fluka™
Catalog # HONCR1162-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    959787-96-3
  • Description:
    (±)-Metoprolol-(isopropyl-d₇) (+)-tartrate salt
  • Melting point:
    110…111 °C
  • Formula:
    C30D14H36N2O6 · C4H6O6
  • Storage temperature:
    2…8 °C
  • Description:
    (±)-Metoprolol-(isopropyl-d₇) (+)-tartrate salt

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results