Order Entry
Denmark
ContactUsLinkComponent
Analytical standard, Reserpine-d₉, reference material, Honeywell Fluka™
Analytical standard, Reserpine-d₉, reference material, Honeywell Fluka™
Catalog # HONCR1000-10MG
CAS Number:  
Analytical standard, Reserpine-d₉, reference material, Honeywell Fluka™
Catalog # HONCR1000-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    84759-11-5
  • Description:
    Reserpine-d₉
  • Melting point:
    273…276 °C
  • Formula:
    C₃₃D₉H₃₁N₂O₉
  • Storage temperature:
    Ambient
  • Description:
    Reserpine-d₉

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results