Order Entry
Denmark
ContactUsLinkComponent
 
Freezer storage box, 100 places, SP Bel-Art
  BELAF188510016
undefined
Freezer storage box, 100 places, SP Bel-Art
  BELAF188510016

 

  • Colour:
    Assorted
  • Description:
    Storage box for 0.5, 1.5 and 2.0 ml tubes and cryo vials
  • Environmentally Preferable:
  • Pk:
    5

 

 

Freezer storage box provide compact, safe storage for tubes and vials.

  • Polypropylene construction
  • Available in a variety of colour choices
  • Hinged lids with snap tabs
  • Alphanumeric labeling

Great for organisation on the bench, in the freezer, or during transport, they can be used at temperatures –80 to +121 °C (112 to +249 °F)

Hinged lids with snap tab open easily and close securely, while clear translucent colour always lets you see inside. Opaque black boxes are ideal for light sensitive samples. Each box position is labeled with an alphanumeric grid for easy management of individual samples. 'No slide' features molded in bottom and lid provide secure stacking.