Contact plates
About this item
Manufactured from transparent PS, these contact plates are for use in the pharmaceutical industry and in hospital environments for determining bacterial contamination of surfaces such as the skin, operating tables, refrigerated gondolas, and work surfaces.
- Aseptic production
- Moulded grid makes it possible to identify the detected contamination per cm² and it facilitates the counting of colonies
- Shape of the lid makes stacking perfectly stable during transport and incubation, and saves space on work surfaces
Ø×H: 65,7×14,7 mm
2 Options Available Below
Product Details & Documents
Manufactured from transparent PS, these contact plates are for use in the pharmaceutical industry and in hospital environments for determining bacterial contamination of surfaces such as the skin, operating tables, refrigerated gondolas, and work surfaces.
- Aseptic production
- Moulded grid makes it possible to identify the detected contamination per cm² and it facilitates the counting of colonies
- Shape of the lid makes stacking perfectly stable during transport and incubation, and saves space on work surfaces
Ø×H: 65,7×14,7 mm
Documents
Product Family Options
Product Information
- ØHeightSterilityPackedAvailabilityPrice
- DELT200208Rodac contact plate
- DELT200218Rodac contact plate