Stem transfection reagents, RmesFect™
Supplier: OZ BIOSCIENCES
Certificates
About this item
RmesFect™ Stem transfection reagent is based on TEE-technology specifically designed for mRNA transfection in stem cells with high efficiency and low toxicity. It is specifically designed for in vitro mRNA (dsRNA) transfection in a large variety stem cells and primary cells.
- Highly efficient, achieve mRNA delivery in all cells
- Ready to use, no need for additional buffer
- Low nucleic acid amount, minimised toxicity
- Protects mRNA against degradation
- Compatible with any culture medium
- Serum compatible, medium change not required
mRNA transfection provides several advantages over plasmid DNA delivery, no need for nuclear uptake - mRNA translation into proteins occurs in the cytoplasm, faster protein expression than DNA transfection, no genomic integration, protein expression in a total promoter independent manner.
Store at –20 °C.
Delivery: Shipped at room temperature.
Specifications
- Cell specific:Stem cells
- mRNA:Yes