DNA transfection reagent, HYPE-5™
Supplier: OZ BIOSCIENCES
Certificates
About this item
HYPE-5™ transfection kit is dedicated to achieve high yield protein expression in mammalian cells. The kit has been designed for maximum recombinant protein expression in HEK293 and CHO cells grown in suspension. It is composed of the HYPE-5™ transfection reagent (a lipofection reagent) and HYPE-5™ Blast reagent (improves protein production in CHO cells). The HYPE-5™ transfection kit is ideal for bioreactor, spinner or flasks.
- High yield recombinant protein production
- Suitable for both HEK293 and CHO cells growing in suspension
- Compatible with any synthetic or regular media used for protein production
- Completely animal origin free
Store HYPE-5™ reagent at –20 °C and HYPE-5™ Blast at +4 °C.
Delivery: Shipped at room temperature.
Specifications
- Bioproduction:Yes
- Cell specific:HEK 293 / CHO suspension
Specifications
Related Information
Frequently Bought Together