Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% (dry basis)
: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
:77-86-1
:
:NH₂C(CH₂OH)₃
:MFCD00004679
:121.14 g/mol
:14,09772
:219…220 °C (10 mmHg)
:171…172 °C
:Ambient
:201-064-4
:GHS07
- Environmentally Preferable:
- Assay (Dry Basis):99.0 - 101.0%
- Assay Absorbance (250 nm, 1 M, 1 cm):≤0.1
- Assay Absorbance (290 nm, 40%, 1 cm):≤0.2
- Loss on Drying:≤1%
- pH (5%, H₂O):10.0 - 11.5
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.1%
- Insoluble Material:≤0.005%
- Heavy Metals (Pb):≤5 ppm
- Iron (Fe):≤5 ppm
- pKa (20 °C):8.30 ±0.15
Frequently Bought Together