Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium azide 99+%

Sodium azide 99+%

Toxic
Health
Sodium azide 99+%
Certificates
Sodium azide 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:14,08581
  • Melting Pt:∼275 °C (dec.)
  • Density:1.846 g/cm³
  • Storage Temperature:Ambient
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Specifications for Product Family

Specification Test Results

  • Form:
    White crystalline powder
  • Assay (Unspecified):
    ≥99.0%
  • Loss on Drying:
    ≤0.5%
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.120,00 kr
    1.120,00 kr
    1 * 100 g
       1.120,00 kr

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1.930,00 kr
    1.930,00 kr
    1 * 500 g
       1.930,00 kr

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 3.950,00 kr
    3.950,00 kr
    1 * 1 kg
       3.950,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.