Collagenase, Type I, from Clostridium histolyticum
Supplier: Thermo Fisher Scientific
Certificates
About this item
Collagenase contains a mixture of collagenase, clostripain, trypsin, caseinase and other lytic enzymes.
- Enzyme activity: ≥125 units/mg powder (dry basis)
- Enzyme activity: ≥0,15 Wunsch units/mg (powder dry basis)
One unit liberates 1 μmol of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5. One unit is equal to 0,0012 Wunsch units.
Specifications
- CAS No.:9001-12-1
- Source:Clostridium histolyticum
- MDL No.:MFCD00130830
- Enzyme name:Collagenase, Type I
Specifications
Frequently Bought Together