(R)-(-)-2-Amino-5-phosphonopentanoic acid ≥95%
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: D-2-Amino-5-phosphonovaleric acid, (R)-2-Amino-5-phosphonopentanoic acid, D-AP5
CAS Number:79055-68-8
:
Formula:C₅H₁₂NO₅P
MDL Number:MFCD00078839
Molecular Weight:197.127 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥95%
Specification Test Results
Frequently Bought Together