Order Entry
Denmark
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ IXSelect
Affinity chromatography columns, HiTrap™ IXSelect
Supplier:   Cytiva
Affinity chromatography columns, HiTrap™ IXSelect
Supplier:  Cytiva
Certificates
Affinity chromatography columns, HiTrap™ IXSelect
2 Options Available Below
Product Details & Documents
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
  • IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Product Family Options
Product Information
  • Column hardware
    Capacity
    Availability
    Price
  • GEHE17-3714-11
    HiTrap™ IXSelect
    HiTrap™ IXSelect
    PP
    1 ml
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 8.280,00 kr
    8.280,00 kr
    1 * 5 Items
       8.280,00 kr
  • GEHE17-3714-12
    HiTrap™ IXSelect
    HiTrap™ IXSelect
    PP
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.240,00 kr
    8.240,00 kr
    1 * 1 Items
       8.240,00 kr