Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-17E (from <i>E. coli</i>)

Mouse Recombinant IL-17E (from E. coli)

Mouse Recombinant IL-17E (from <i>E. coli</i>)
Certificates
Mouse Recombinant IL-17E (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT200-60-5UG
  • SHBT200-60-25UG
  • SHBT200-60-100UG
  • SHBT200-60-500UG
  • SHBT200-60-1MG
  • Protein/peptide name:
  • IL-17E
  • IL-17E
  • IL-17E
  • IL-17E
  • IL-17E
  • Protein synonyms:
  • IL-25
  • IL-25
  • IL-25
  • IL-25
  • IL-25
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q8VHC9
  • Q8VHC9
  • Q8VHC9
  • Q8VHC9
  • Q8VHC9
  • Sequence:
  • MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥90%
  • ≥90%
  • ≥90%
  • ≥90%
  • ≥90%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • Molecular weight:
  • 17,6/35,2 kDa
  • 17,6/35,2 kDa
  • 17,6/35,2 kDa
  • 17,6/35,2 kDa
  • 17,6/35,2 kDa
  • Reconstitution instructions:
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Sterile 10 mM HCl at 0,1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 574,00 kr
    574,00 kr
    1 * 1 Each
       574,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-60-5UG
    Protein/peptide name
    IL-17E
    Protein synonyms
    IL-25
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8VHC9
    Sequence
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    17,6/35,2 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    25 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-60-25UG
    Protein/peptide name
    IL-17E
    Protein synonyms
    IL-25
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8VHC9
    Sequence
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    17,6/35,2 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.570,00 kr
    3.570,00 kr
    1 * 1 Each
       3.570,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-60-100UG
    Protein/peptide name
    IL-17E
    Protein synonyms
    IL-25
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8VHC9
    Sequence
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    17,6/35,2 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 15.400,00 kr
    15.400,00 kr
    1 * 1 Each
       15.400,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-60-500UG
    Protein/peptide name
    IL-17E
    Protein synonyms
    IL-25
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8VHC9
    Sequence
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    17,6/35,2 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 25.640,00 kr
    25.640,00 kr
    1 * 1 mg
       25.640,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-60-1MG
    Protein/peptide name
    IL-17E
    Protein synonyms
    IL-25
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8VHC9
    Sequence
    MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥90%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    17,6/35,2 kDa
    Reconstitution instructions
    Sterile 10 mM HCl at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.