Mouse Recombinant IL-22 (from E. coli)
About this item
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains. IL-22 is a potent mediator of cellular inflammatory responses and wound healing.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
4 Options Available Below
Product Details & Documents
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains. IL-22 is a potent mediator of cellular inflammatory responses and wound healing.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT200-77-10UG
- SHBT200-77-100UG
- SHBT200-77-500UG
- SHBT200-77-1MG
- Protein/peptide name:
- IL-22
- IL-22
- IL-22
- IL-22
- Protein synonyms:
- IL-TIF
- IL-TIF
- IL-TIF
- IL-TIF
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q9JJY9
- Q9JJY9
- Q9JJY9
- Q9JJY9
- Sequence:
- MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Form:
- Lyophilised
- Lyophilised
- Lyophilised
- Lyophilised
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 16,8 kDa
- 16,8 kDa
- 16,8 kDa
- 16,8 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT200-77-10UGProtein/peptide nameIL-22Protein synonymsIL-TIFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9JJY9SequenceMLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACVProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight16,8 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-77-100UGProtein/peptide nameIL-22Protein synonymsIL-TIFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9JJY9SequenceMLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACVProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight16,8 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-77-500UGProtein/peptide nameIL-22Protein synonymsIL-TIFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9JJY9SequenceMLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACVProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight16,8 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT200-77-1MGProtein/peptide nameIL-22Protein synonymsIL-TIFProtein/peptide typeRecombinantProtein functionCytokineSpeciesMouseSourceE. coliConjugationUnconjugatedGene IDQ9JJY9SequenceMLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACVProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormLyophilisedFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight16,8 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



