Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant IL-33 (from <i>E. coli</i>)

Human Recombinant IL-33 (from E. coli)

Human Recombinant IL-33 (from <i>E. coli</i>)
Certificates
Human Recombinant IL-33 (from <i>E. coli</i>)

4 Options Available Below

Product Details & Documents

Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle and airway epithelial cells. IL-33 signals through the interleukin 1 receptor-like 1 (IL-1R1) and interleukin-1 receptor accessory protein (IL1RAP) receptors to ativate NF-κB and MAPK signaling pathways. IL-33 functions to induce type 2 cytokine production in polarized Type 2 helper T (Th2) cells.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-145-10UG
  • SHBT100-145-100UG
  • SHBT100-145-500UG
  • SHBT100-145-1MG
  • Protein/peptide name:
  • IL-33
  • IL-33
  • IL-33
  • IL-33
  • Protein synonyms:
  • NP-HEV
  • NP-HEV
  • NP-HEV
  • NP-HEV
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • O95760
  • O95760
  • O95760
  • O95760
  • Sequence:
  • MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
  • MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
  • MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
  • MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • 10 mM sodium phosphate, pH 7,5
  • Molecular weight:
  • 18,1 kDa
  • 18,1 kDa
  • 18,1 kDa
  • 18,1 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-145-10UG
    Protein/peptide name
    IL-33
    Protein synonyms
    NP-HEV
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O95760
    Sequence
    MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    18,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 5.510,00 kr
    5.510,00 kr
    1 * 1 Each
       5.510,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-145-100UG
    Protein/peptide name
    IL-33
    Protein synonyms
    NP-HEV
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O95760
    Sequence
    MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    18,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 27.350,00 kr
    27.350,00 kr
    1 * 1 Each
       27.350,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-145-500UG
    Protein/peptide name
    IL-33
    Protein synonyms
    NP-HEV
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O95760
    Sequence
    MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    18,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 45.590,00 kr
    45.590,00 kr
    1 * 1 mg
       45.590,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-145-1MG
    Protein/peptide name
    IL-33
    Protein synonyms
    NP-HEV
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O95760
    Sequence
    MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: D10S cell proliferation, ED50 ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium phosphate, pH 7,5
    Molecular weight
    18,1 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.