Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Mouse Recombinant GDNF (from E. coli)

Mouse Recombinant GDNF (from <i>E. coli</i>)
 
Mouse Recombinant GDNF (from <i>E. coli</i>)
Mouse Recombinant GDNF (from E. coli)

 

 

Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.

  • High quality
  • Low endotoxin
  • Affordable

 : For research use only.

 

 

 

  • Catalog No:
  • SHBT200-37-10UG
  • SHBT200-37-100UG
  • SHBT200-37-500UG
  • SHBT200-37-1MG
  • Protein/peptide name:
  • GDNF
  • GDNF
  • GDNF
  • GDNF
  • Protein synonyms:
  • ATF-1
  • ATF-1
  • ATF-1
  • ATF-1
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P48540
  • P48540
  • P48540
  • P48540
  • Sequence:
  • MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • Molecular weight:
  • 15,1/30,2 kDa
  • 15,1/30,2 kDa
  • 15,1/30,2 kDa
  • 15,1/30,2 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT

 

Product Family Options

 

  • Pack type
    Pk
     
     
  • Vial
    10 µG
       
       
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

     
     
    Specifications
    Cat. No.
    SHBT200-37-10UG
    Protein/peptide name
    GDNF
    Protein synonyms
    ATF-1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P48540
    Sequence
    MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    15,1/30,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
       
       
    1 * 1 Each
    1 * 1 Each - 5.510,00 kr
    5.510,00 kr
    1 * 1 Each
       5.510,00 kr

     
     
    Specifications
    Cat. No.
    SHBT200-37-100UG
    Protein/peptide name
    GDNF
    Protein synonyms
    ATF-1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P48540
    Sequence
    MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    15,1/30,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
       
       
    1 * 1 Each
    1 * 1 Each - 27.350,00 kr
    27.350,00 kr
    1 * 1 Each
       27.350,00 kr

     
     
    Specifications
    Cat. No.
    SHBT200-37-500UG
    Protein/peptide name
    GDNF
    Protein synonyms
    ATF-1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P48540
    Sequence
    MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    15,1/30,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
       
       
    1 * 1 mg
    1 * 1 mg - 45.590,00 kr
    45.590,00 kr
    1 * 1 mg
       45.590,00 kr

     
     
    Specifications
    Cat. No.
    SHBT200-37-1MG
    Protein/peptide name
    GDNF
    Protein synonyms
    ATF-1
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P48540
    Sequence
    MSPDKQAAL PRRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    15,1/30,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT