Rat Recombinant IGF-I (from E. coli)
About this item
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
5 Options Available Below
Product Details & Documents
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT300-01-10UG
- SHBT300-01-50UG
- SHBT300-01-100UG
- SHBT300-01-500UG
- SHBT300-01-1MG
- Protein/peptide name:
- IGF-I
- IGF-I
- IGF-I
- IGF-I
- IGF-I
- Protein synonyms:
- mechano growth factor|IGF-IA|Somatomedin C
- mechano growth factor|IGF-IA|Somatomedin C
- mechano growth factor|IGF-IA|Somatomedin C
- mechano growth factor|IGF-IA|Somatomedin C
- mechano growth factor|IGF-IA|Somatomedin C
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Rat
- Rat
- Rat
- Rat
- Rat
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P08025
- P08025
- P08025
- P08025
- P08025
- Sequence:
- GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
- GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
- GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
- GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
- GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- UniProtKB:
- P08025
- P08025
- P08025
- P08025
- P08025
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 7,7 kDa
- 7,7 kDa
- 7,7 kDa
- 7,7 kDa
- 7,7 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT300-01-10UGProtein/peptide nameIGF-IProtein synonymsmechano growth factor|IGF-IA|Somatomedin CProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP08025SequenceGPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP08025Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight7,7 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT300-01-50UGProtein/peptide nameIGF-IProtein synonymsmechano growth factor|IGF-IA|Somatomedin CProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP08025SequenceGPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP08025Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight7,7 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT300-01-100UGProtein/peptide nameIGF-IProtein synonymsmechano growth factor|IGF-IA|Somatomedin CProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP08025SequenceGPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP08025Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight7,7 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT300-01-500UGProtein/peptide nameIGF-IProtein synonymsmechano growth factor|IGF-IA|Somatomedin CProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP08025SequenceGPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP08025Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight7,7 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT300-01-1MGProtein/peptide nameIGF-IProtein synonymsmechano growth factor|IGF-IA|Somatomedin CProtein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP08025SequenceGPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBP08025Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight7,7 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



