Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant MIP-4 (from E. coli)

Human Recombinant MIP-4 (from <i>E. coli</i>)
Certificates
Human Recombinant MIP-4 (from <i>E. coli</i>)

4 Options Available Below

Product Details & Documents

Macrophage inflammatory protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta, and bone marrow. MIP-4 receptors include the chemokine receptor 8 (CCR8), the G protein-coupled receptor 30 (GPR30), and the phosphatidylinositol transfer protein membrane-associate 3 (PITPNM3). MIP-4 acts as a chemoattractant for naive T cells, CD4+ T cells, CD8+ T cells, and nonactivated lymphocytes. Further, MIP-4 promotes breast cancer metastasis and attenuates the activation of acute lymphocytic leukemia B cells.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

 : For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-136-10UG
  • SHBT100-136-100UG
  • SHBT100-136-500UG
  • SHBT100-136-1MG
  • Protein/peptide name:
  • MIP-4
  • MIP-4
  • MIP-4
  • MIP-4
  • Protein synonyms:
  • DC-CK1|CCL18|AMAC-1|PARC
  • DC-CK1|CCL18|AMAC-1|PARC
  • DC-CK1|CCL18|AMAC-1|PARC
  • DC-CK1|CCL18|AMAC-1|PARC
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P55774
  • P55774
  • P13501
  • P55774
  • Sequence:
  • AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
  • AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
  • AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
  • AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • 0,1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 7,9 kDa
  • 7,9 kDa
  • 7,9 kDa
  • 7,9 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    10 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-136-10UG
    Protein/peptide name
    MIP-4
    Protein synonyms
    DC-CK1|CCL18|AMAC-1|PARC
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P55774
    Sequence
    AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    7,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.570,00 kr
    3.570,00 kr
    1 * 1 Each
       3.570,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-136-100UG
    Protein/peptide name
    MIP-4
    Protein synonyms
    DC-CK1|CCL18|AMAC-1|PARC
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P55774
    Sequence
    AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    7,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 15.400,00 kr
    15.400,00 kr
    1 * 1 Each
       15.400,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-136-500UG
    Protein/peptide name
    MIP-4
    Protein synonyms
    DC-CK1|CCL18|AMAC-1|PARC
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P13501
    Sequence
    AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    7,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 25.640,00 kr
    25.640,00 kr
    1 * 1 mg
       25.640,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-136-1MG
    Protein/peptide name
    MIP-4
    Protein synonyms
    DC-CK1|CCL18|AMAC-1|PARC
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P55774
    Sequence
    AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic Acid (TFA)
    Molecular weight
    7,9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT