Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-2 (from <i>E. coli</i>)

Mouse Recombinant IL-2 (from E. coli)

Mouse Recombinant IL-2 (from <i>E. coli</i>)
Certificates
Mouse Recombinant IL-2 (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

Caution: For research use only.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT200-17-5UG
  • SHBT200-17-20UG
  • SHBT200-17-100UG
  • SHBT200-17-500UG
  • SHBT200-17-1MG
  • Protein/peptide name:
  • IL-2
  • IL-2
  • IL-2
  • IL-2
  • IL-2
  • Protein synonyms:
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Aldesleukin|TCGF|T cell growth factor
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P04351
  • P04351
  • P04351
  • P04351
  • P04351
  • Sequence:
  • MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
  • MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
  • MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
  • MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
  • MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM sodium citrate, pH 4,0
  • 10 mM sodium citrate, pH 4,0
  • 10 mM sodium citrate, pH 4,0
  • 10 mM sodium citrate, pH 4,0
  • 10 mM sodium citrate, pH 4,0
  • Molecular weight:
  • 17,4 kDa
  • 17,4 kDa
  • 17,4 kDa
  • 17,4 kDa
  • 17,4 kDa
  • Reconstitution instructions:
  • Sterile 10 mM acetic acid at 0.1 mg/ml
  • Sterile 10 mM acetic acid at 0.1 mg/ml
  • Sterile 10 mM acetic acid at 0.1 mg/ml
  • Sterile 10 mM acetic acid at 0.1 mg/ml
  • Sterile 10 mM acetic acid at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 574,00 kr
    574,00 kr
    1 * 1 Each
       574,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-17-5UG
    Protein/peptide name
    IL-2
    Protein synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04351
    Sequence
    MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 4,0
    Molecular weight
    17,4 kDa
    Reconstitution instructions
    Sterile 10 mM acetic acid at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    20 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-17-20UG
    Protein/peptide name
    IL-2
    Protein synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04351
    Sequence
    MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 4,0
    Molecular weight
    17,4 kDa
    Reconstitution instructions
    Sterile 10 mM acetic acid at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.570,00 kr
    3.570,00 kr
    1 * 1 Each
       3.570,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-17-100UG
    Protein/peptide name
    IL-2
    Protein synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04351
    Sequence
    MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 4,0
    Molecular weight
    17,4 kDa
    Reconstitution instructions
    Sterile 10 mM acetic acid at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 14.260,00 kr
    14.260,00 kr
    1 * 1 Each
       14.260,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-17-500UG
    Protein/peptide name
    IL-2
    Protein synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04351
    Sequence
    MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 4,0
    Molecular weight
    17,4 kDa
    Reconstitution instructions
    Sterile 10 mM acetic acid at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 23.750,00 kr
    23.750,00 kr
    1 * 1 mg
       23.750,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT200-17-1MG
    Protein/peptide name
    IL-2
    Protein synonyms
    Aldesleukin|TCGF|T cell growth factor
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P04351
    Sequence
    MAPTSSSTSS STAEAQQQQQ QQQQQQQHLE QLLMDLQELL SRMENYRNLK LPRMLTFKFY LPKQATELKD LQCLEDELGP LRHVLDLTQS KSFQLEDAEN FISNIRVTVV KLKGSDNTFE CQFDDESATV VDFLRRWIAF CQSIISTSPQ
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM sodium citrate, pH 4,0
    Molecular weight
    17,4 kDa
    Reconstitution instructions
    Sterile 10 mM acetic acid at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.