VWR®, Bottle carrying systems
Leverandør: DWK Life Sciences
Certifikater
Om denne vare
This modular bottle carrying system fits standard laboratory bottles that have a GL 45 neck finish.
- Compatible with four bottle sizes 2, 5, 10 and 20 litres
- All materials are fully autoclavable
- Makes pouring liquids from the larger bottles more comfortable and safer
It is ergonomically designed and offers health and safety benefits to the user. It allows easy and safe transport of bottles around the laboratory or production facility.
Relateret information
Frequently Bought Together