Guanine
Supplier: RESEARCH ORGANICS
Jump to:
About this item
Synonyms: 2-Amino-1H-purin-6(7H)-one
- CAS Number:73-40-5
- Formula:C₅H₅N₅O
- MDL Number:MFCD00071533
- Molecular Weight:151.13 g/mol
- Merck Index:13,04580
- Melting Pt:> 220 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-799-8
2 Options Available Below
Product Details & Documents
Synonyms: 2-Amino-1H-purin-6(7H)-one
- CAS Number:73-40-5
- Formula:C₅H₅N₅O
- MDL Number:MFCD00071533
- Molecular Weight:151.13 g/mol
- Merck Index:13,04580
- Melting Pt:> 220 °C
- Density:1.67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-799-8
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



