Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant FGF-10 (from <i>E. coli</i>)

Human Recombinant FGF-10 (from E. coli)

Human Recombinant FGF-10 (from <i>E. coli</i>)
Certificates
Human Recombinant FGF-10 (from <i>E. coli</i>)

5 Options Available Below

Product Details & Documents

Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis.FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinising epidermal cells, and induces the migration and invasion of cancer cells.

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-183-5UG
  • SHBT100-183-25UG
  • SHBT100-183-100UG
  • SHBT100-183-500UG
  • SHBT100-183-1MG
  • Protein/peptide name:
  • FGF-10
  • FGF-10
  • FGF-10
  • FGF-10
  • FGF-10
  • Protein synonyms:
  • Keratinocyte growth factor 2
  • Keratinocyte growth factor 2
  • Keratinocyte growth factor 2
  • Keratinocyte growth factor 2
  • Keratinocyte growth factor 2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • O15520
  • O15520
  • O15520
  • O15520
  • O15520
  • Sequence:
  • MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNNDCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
  • MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
  • MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
  • MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
  • MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
  • Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
  • Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
  • Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
  • Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 10 mM Sodium phosphate, pH 7,5
  • 10 mM Sodium phosphate, pH 7,5
  • 10 mM Sodium phosphate, pH 7,5
  • 10 mM Sodium phosphate, pH 7,5
  • 10 mM Sodium phosphate, pH 7,5
  • Molecular weight:
  • 19,3 kDa
  • 19,3 kDa
  • 19,3 kDa
  • 19,3 kDa
  • 19,3 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 574,00 kr
    574,00 kr
    1 * 1 Each
       574,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-183-5UG
    Protein/peptide name
    FGF-10
    Protein synonyms
    Keratinocyte growth factor 2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O15520
    Sequence
    MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNNDCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, pH 7,5
    Molecular weight
    19,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    25 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.430,00 kr
    1.430,00 kr
    1 * 1 Each
       1.430,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-183-25UG
    Protein/peptide name
    FGF-10
    Protein synonyms
    Keratinocyte growth factor 2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O15520
    Sequence
    MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, pH 7,5
    Molecular weight
    19,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.570,00 kr
    3.570,00 kr
    1 * 1 Each
       3.570,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-183-100UG
    Protein/peptide name
    FGF-10
    Protein synonyms
    Keratinocyte growth factor 2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O15520
    Sequence
    MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, pH 7,5
    Molecular weight
    19,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 15.400,00 kr
    15.400,00 kr
    1 * 1 Each
       15.400,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-183-500UG
    Protein/peptide name
    FGF-10
    Protein synonyms
    Keratinocyte growth factor 2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O15520
    Sequence
    MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, pH 7,5
    Molecular weight
    19,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 25.640,00 kr
    25.640,00 kr
    1 * 1 mg
       25.640,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-183-1MG
    Protein/peptide name
    FGF-10
    Protein synonyms
    Keratinocyte growth factor 2
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    O15520
    Sequence
    MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEIGVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    10 mM Sodium phosphate, pH 7,5
    Molecular weight
    19,3 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay