Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant EGF (from <i>E. coli</i>)

Human Recombinant EGF (from E. coli)

Human Recombinant EGF (from <i>E. coli</i>)
Certificates
Human Recombinant EGF (from <i>E. coli</i>)

3 Options Available Below

Product Details & Documents

Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.

  • High quality
  • Low endotoxin
  • Affordable

Caution: This product is for research use only and is not for use in humans.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SHBT100-26AF-5UG
  • SHBT100-26AF-100UG
  • SHBT100-26AF-500UG
  • Protein/peptide name:
  • EGF (AF)
  • EGF (AF)
  • EGF (AF)
  • Protein synonyms:
  • Urogastrone|URG
  • Urogastrone|URG
  • Urogastrone|URG
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P01133
  • P01133
  • P01133
  • Sequence:
  • NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
  • NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
  • NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
  • Protease-free:
  • N
  • N
  • N
  • Animal-free:
  • Y
  • Y
  • Y
  • Nuclease-free:
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
  • Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
  • Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
  • Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • 0,1% Trifluoroacetic acid (TFA)
  • Molecular weight:
  • 6,2 kDa
  • 6,2 kDa
  • 6,2 kDa
  • Reconstitution instructions:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • Tested applications:
  • Bioassay
  • Bioassay
  • Bioassay

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Vial
    5 µG
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 10.450,00 kr
    10.450,00 kr
    1 * 5 mg
       10.450,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-26AF-5UG
    Protein/peptide name
    EGF (AF)
    Protein synonyms
    Urogastrone|URG
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01133
    Sequence
    NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    6,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    100 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 618,00 kr
    618,00 kr
    1 * 1 Each
       618,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-26AF-100UG
    Protein/peptide name
    EGF (AF)
    Protein synonyms
    Urogastrone|URG
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01133
    Sequence
    NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    6,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay
  • Vial
    500 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.570,00 kr
    1.570,00 kr
    1 * 1 Each
       1.570,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    SHBT100-26AF-500UG
    Protein/peptide name
    EGF (AF)
    Protein synonyms
    Urogastrone|URG
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P01133
    Sequence
    NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
    Protease-free
    N
    Animal-free
    Y
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0,1
    Biological activity
    Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0,1% Trifluoroacetic acid (TFA)
    Molecular weight
    6,2 kDa
    Reconstitution instructions
    Sterile water at 0,1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
    Tested applications
    Bioassay